Learn More
Aviva Systems Biology Corp SHARPIN Antibody - C-terminal region (ARP59390_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59390_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SHARPIN. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SHARPIN. Alternate names: DKFZp434N1923, SIPL1ApplicationWB. Species reactivity: Cow, Human, Rat
Catalog No. 50-286-680