Learn More
American Research Products Inc Sheep C reactiVe protein CRP
Supplier:  American Research Products Inc CSBE15081SH
Encompass
Sheep C-reactiVe protein (CRP) ELISA Kit from CUSABIO Detection Range 0 625 g/mL-40 g/mL Sensitivity 0 156 g/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28581