Learn More
Grainger Shelf Cap 700 lb 48 x 24 in
Supplier:  Grainger 14C732
Encompass_Preferred
The Hallowell Shelf is a 48 inch wide by 24 inch deep steel shelf with a weight capacity of 800 lbs. It has a smooth finish and is made of 22 gauge steel with return edges for strength and safety. The shelf has mounting holes for easy installation and can be used for industrial storage and material handling applications. With its sturdy steel construction, the Hallowell shelf provides a durable storage solution for warehouses, factories, garages, and other commercial settings.
Catalog No. 50-275-2677