Learn More
Grainger Shelf Cap 755 lb 48 x 36 in
Supplier:  Grainger 14C673
Encompass_Preferred
The Hallowell Shelf is a 48 inch wide by 36 inch deep steel shelf with a weight capacity of 800 lbs. It has a smooth finish and is made of 22 gauge steel with return edges for strength and safety. The shelf has mounting holes for easy installation and can be used for industrial storage and material handling applications. With its heavy duty steel construction, the Hallowell shelf provides sturdy and reliable storage space.
Catalog No. 50-275-3630