Learn More
Bellco Glass, Inc. Shell Amber Glass Vial,1.0mL 8x40mm
SDP
Supplier:  Bellco Glass, Inc. 207000120
Encompass_Preferred
Designed for use with the water wisp 96-position auto sampler plug insert. MADE IN THE USA. Comes in amber and clear vials Size: 8x40mm Plug insert sold separately
Catalog No. 50-194-5366