Learn More
Medchemexpress LLC Shellac (wax-free) | 9000-59-3 | MFCD03101446 | Mixture - no single molecular weight | Mixture - no single chemical formula | 100 G
SDP
Supplier:  Medchemexpress LLC HYW133969100G
Encompass_Preferred
Shellac (wax-free) is a natural, biodegradable resin obtained from the secretions of the lac insect (Kerria lacca). Supplied as a wax-free resin, it functions as a film former, binder, and encapsulating agent for coatings, pharmaceutical and food supplement formulations, and cosmetic applications. Composition is variable and referenced under CAS 9000-59-3.
  • Biodegradable natural resin suitable for environmentally sensitive applications.
  • Wax-free formulation for cleaner processing and consistent performance.
  • Functions as a film former and wood coating.
  • Provides binding and encapsulation for pharmaceuticals and supplements.
  • Compatible with cosmetic formulations as a binder or coating.
  • Available in laboratory package sizes for formulation and testing.
Catalog No. 50-002-83357