Learn More
Mitegen Shelved Puck Shipping Cane Kit, Holds 7 UniPucks, ALS or Cryo-EM Pucks, For Use With The CX100 Dryshipper, Incl. Locking Rod & Hooked Handle
SDP
Supplier:  Mitegen MCP111065
Encompass
This Shelved Puck Shipping Cane Kit provides a secure and organized solution for the transport and storage of cryogenic sample pucks. Designed for use in dryshippers, including models like the CX100, this cane is compatible with Universal V-1 Pucks, ALS Pucks, and Cryo-EM Pucks. Each cane is capable of holding up to seven pucks, ensuring efficient utilization of space and systematic sample management. The kit includes the shelved shipping cane itself, a crucial locking rod for enhanced security during transport, and a convenient hooked handle for easy manipulation. It is imperative to always insert the locking rod when shipping pucks in dry-shipping dewars to maintain sample integrity. Furthermore, this versatile cane can be utilized as a storage cane in standard storage dewars with a compatible S-Hook Cane Storage Adapter, offering flexibility for your laboratory's cryogenic needs.
Catalog No. 50-118-7201