Learn More
Mitegen Shelved Storage Cane for MiTeGen Generation 2 Cryo-EM Pucks - Holds up to 10 Pucks, Compatible with IC Biomedical HC35, TW35-10 and ICB35-10
SDP
Supplier:  Mitegen MCEMSTC235
Encompass_Preferred
The MiTeGen shelved storage cane is designed to hold the MiTeGen 2nd Generation Cryo-EM Pucks SKU M-CEMG2-1. It holds up to 10 pucks and is compatible with IC Biomedical HC35, TW35-10 and ICB35-10 storage dewars. Features many safety and quality of life improvements, such as steel shelves which engage the magnets on the Cryo-EM pucks which help avoid pucks falling out while the locking rod is not engaged.
Catalog No. 50-004-94992