Learn More
Medchemexpress LLC HY-112210 5mg Medchemexpress, Shield-1 CAS:914805-33-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1122105MG
Encompass_Preferred
Medchemexpress, HY-112210 5mg Shield-1 CAS:914805-33-7 Shield-1 is a specific, cell-permeant and high-affinity ligand of FK506-binding protein-12 (FKBP), and reverses the instability by binding to mutated FKBP (mtFKBP), allowing conditional expression of mtFKBP-fused proteins. Shield-1 can stabilize the entire fusion protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9062