Learn More
Enzo Life Sciences Shikonin (10mg). CAS: 517-89-5
SDP
Supplier:  Enzo Life Sciences BMLCT1150010
Encompass_Preferred
Shikonin inhibits TNF-α- and B6 melanoma-induced angiogenesis in mice. It blocks expression of integrin αvβ3 and inhibits endothelial cell proliferation and migration in vitro. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: +4°C.
Catalog No. 50-201-0483