Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-SHIN1 | 2444764-09-2 | 99.1% | 400.47 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY112066B50MG

Encompass_Preferred

(-)-SHIN1 ((-)-RZ-2994) is an inactive (-) enantiomer of SHIN1, intended for research use only. It is not for medical applications or sale to patients.

  • Appearance: Solid, white to off-white.
  • Identified uses: Laboratory chemicals and substance manufacturing.
  • Recommended storage: 4°C under nitrogen.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (under nitrogen).
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Target: Drug isomer.

Catalog No. 50-004-37218


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.