Learn More
Medchemexpress LLC (+)-SHIN1 ((+)-RZ-2994) | 2443966-90-1 | 99.6% | 400.47 | 10 MG

Supplier: Medchemexpress LLC HY112066A10MG
(+)-SHIN1 ((+)-RZ-2994) is an active (+) enantiomer of SHIN1. It is potent against cytosolic SHMT1 with an IC50 for blocking growth of less than 50 nM and 870 nM in SHMT2 deletion cells and HCT-116 cells, respectively.
- Purity: 99.64%
- Cas number: 2443966-90-1
- Molecular weight: 400.47
- Formula: C24H24N4O2
- Appearance: Solid
- Color: White to off-white
- Solubility: ≥ 100 mg/mL (249.71 mM) in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.