Learn More
Medchemexpress LLC (+)-SHIN1 ((+)-RZ-2994) | 2443966-90-1 | 99.64% | 400.47 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY112066A50MG
Encompass_Preferred
(+)-SHIN1 ((+)-RZ-2994) is an active (+) enantiomer of SHIN1. It is potent against cytosolic SHMT1 with an IC50 for blocking growth of less than 50 nM and 870 nM in SHMT2 deletion cells and HCT-116 cells, respectively. It is for research use only.
  • Active (+) enantiomer of SHIN1.
  • Potent against cytosolic SHMT1.
  • For research use only.
Catalog No. 50-004-39038