Learn More
Scientific Sales Cardinal Health, 405M, Surgical Mask with Ties, Blue
SDP
Supplier:  Scientific Sales 405M
Encompass
The Cardinal Health 405M Surgical Mask is a level 3 mask that provides >98% bacterial filtration efficiency. It has a fluid resistant 160 mmHg pressure differential and a soft inner facing to provide comfort. The mask's ear loops are ultrasonically bonded to the mask material. It is latex-free and FDA cleared for use as a surgical mask. The 405M mask provides a high level of protection for procedures with moderate to heavy amounts of fluid, spray and/or aerosols.
Catalog No. NC1682652