Learn More
Waters Corp Waters Corporation Shodex KD-802.5 GPC Column, 5 μm, 8 × 300 mm, DMF, 20,000 MW Exclusion Limit
Supplier: Waters Corp WAT034118
The Shodex KD-802.5 GPC Column is an ultra-high-efficiency column engineered for high-resolution gel permeation chromatography (GPC) applications. It is constructed with 5 μm styrene divinylbenzene resin particles and measures 8 × 300 mm. The column comes packed in Dimethylformamide (DMF) and offers a Polystyrene Exclusion Limit of 20,000. This column is suitable for precise molecular weight determination of organic-soluble polymers in DMF.
- Particle size: 5 μm
- Dimensions: 8 × 300 mm
- Material: Styrene divinylbenzene resins
- Packed solvent: Dimethylformamide (DMF)
- Polystyrene exclusion limit: 20,000
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.