Learn More
Waters Corp Waters Guard Column, 5 μm, 4.6 mm ID, 10 mm Length, Polymer
Supplier:  Waters Corp WAT034106
Encompass_Preferred
The Shodex KF-G Guard Column is designed to maintain analytical column performance by extending its lifespan, primarily supporting legacy methods. It removes chemical and particulate materials from the mobile phase, protecting the analytical column from harm and preventing impurities and interference in analysis. This guard column supports a broad range of applications for low to high molecular weight compounds, aiding in characterizing polymer molecular weight distribution to determine polymer physical properties.
  • Not intended for improving column resolving power.
  • Easy to use and attached with analytical columns.
  • Offers a broad pH range from 2 to 12 and sustains a maximum pressure of 6000 psi (415 Bar).
Catalog No. 50-785-323