Learn More
Supply Solutions Shodex - SUGAR SP0810. Separates saccharides in SEC+Ligand exchange mode. 8.0x300, 7 um. Sulfo (Pb2+) ligand.
Supplier:  Supply Solutions SHODEX F6378105
Encompass_Preferred
Shodex - SUGAR SP0810. Separates saccharides in SEC+Ligand exchange mode. 8.0x300, 7 um. Sulfo (Pb2+) ligand.
Catalog No. NC1422005