Learn More
Medchemexpress LLC Shogaol | 555-66-8 | 99.84% | 276.37 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1461610MM/1ML
Encompass_Preferred
Shogaol is an active compound derived from Ginger (Zingiber officinale Rosc) and demonstrates a range of biological activities, including anticancer, anti-inflammatory, and antioxidant effects.
  • Exhibits anticancer activity across various cell lines.
  • Inhibits LPS-induced nitric oxide production, demonstrating anti-inflammatory action.
  • Upregulates antioxidant defense systems in cells and in vivo.
  • Protects against liver damage and lipid peroxidation.
  • Restores hepatic antioxidant enzymes.
  • Stimulates phosphorylation of mitogen-activated protein kinases (MAPKs).
  • Offers neuroprotection against beta-Amyloid insult.
  • Modulates human TRPV1 and rat TRPA1 ion channels.
Catalog No. 50-003-59842