Learn More
Perkin Elmer US LLC Short Graphite Ferrules - 0.0625 in x 0.4 mm I.D./0.5 mm I.D., Pkg. 10
Supplier:  Perkin Elmer US LLC 09200785
Encompass_Preferred
Of the three ferrule types available, graphite is the softest, making it less damaging to capillary columns when overtightened. Graphite seals easily and does not stick to glass columns. These one-piece ferrules do not require rear ferrules. Quantity: Package of 10 Nut Size: 1/16 inch Column Inner Diameter: 0.32 mm Ferrule Inner Diameter: 0.4 mm, 0.5 mm
Catalog No. 50-210-4379