Learn More
Sklar Instruments Shortbent Suture Scissors, Sklar, 3-1/2 in., Blunt/Blunt, Curved, Smooth, Shortbent, Stainless Steel. EA/1.
SDP
Supplier:  Sklar Instruments 222935
Encompass_Preferred
Shortbent Suture Scissors, Sklar, 3-1/2 in., Blunt/Blunt, Curved, Smooth, Shortbent, Stainless Steel. Sklar Shortbent Suture Scissors are primarily used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them slightly before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Shortbent suture scissors are smaller than many standard suture scissors and they have delicate blades, ideal for removal of small sutures. They should not be used for cutting tissue. This product has straight shanks with smooth, curved blades, blunt/blunt tips and a length of 3-1/2 inches.
Catalog No. 50-143-7862