Learn More
Bellco Glass, Inc. Shortie Pipet, 2x.1mL Cotton Plug Top
SDP
Supplier:  Bellco Glass, Inc. 122602010
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Shorter length facilitates use under biological hood. MADE IN THE USA Case qty-18 TOLERANCE +-(ML):0.025OAL (MM):175COLOR CODE: Green Graduation: 0.1
Catalog No. 50-194-4636