Learn More
Bellco Glass, Inc. Shorty Cotton Plug Top Pipette, 1x.1mL-Bulk
SDP
Supplier:  Bellco Glass, Inc. 120601010
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Shorter length facilitates use use under biological hoods. MADE IN THE USA Case Qty: 150 TOLERANCE +-(ML):0.02OAL (MM):175COLOR CODE: Red Graduation: 0.1
Catalog No. 50-194-4583