Learn More
Selleck Chemical LLC SHP099 HCl
SDP
Supplier:  Selleck Chemical LLC S827810MM/1ML
Encompass_Preferred
SHP099 is a highly potent selective and orally bioavailable small-molecule SHP2 inhibitor with an IC50 value of 0 071 M and shows no activity against SHP1
Catalog No. 50-000-01522