Learn More
Medchemexpress LLC SHP389 10mg | 2235394-90-6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11445310MG
Encompass_Preferred
SHP389 is an allosteric SHP2 inhibitor that blocks SHP2 enzymatic activity and downstream ERK phosphorylation with an IC50 of 36 nM. It is supplied as a white to off-white solid with high reported purity and is intended for research use.
  • Inhibits SHP2 and p-ERK with IC50 of 36 nM.
  • High purity suitable for biochemical and cell-based assays.
  • Supplied as a stable solid with recommended cold storage.
  • Provided with analytical documents such as COA, SDS, and LCMS for quality verification.
  • Available in multiple pack sizes and solution formats for assay flexibility.
Catalog No. 50-002-28093