Learn More
Medchemexpress LLC SHP389 1mg | 2235394-90-6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1144531MG
Encompass_Preferred
SHP389 is an allosteric SHP2 inhibitor for research use. It inhibits SHP2 and p-ERK with an IC50 of 36 nM and shows antiproliferative activity in Detroit 562 cells (IC50 4.13 μM, 24 h).
  • Allosteric SHP2 inhibition, IC50 36 nM for SHP2 and p-ERK.
  • Demonstrated antiproliferative activity in Detroit 562 cells (IC50 4.13 μM, 24 h).
  • High purity suitable for biochemical and cellular studies.
  • Solid, white to off-white appearance for easy handling.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-26702