Learn More
Medchemexpress LLC SHP389 5mg | 2235394-90-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1144535MG
Encompass_Preferred
SHP389 is a small-molecule allosteric inhibitor of the SHP2 (PTPN11) phosphatase that inhibits SHP2 enzymatic activity and downstream p-ERK signaling. It is intended for biochemical and cellular research and supplied as a small solid quantity suitable for laboratory use.
  • Allosteric inhibition of SHP2 with reported IC50 36 nM
  • Inhibits p-ERK signaling in cellular assays
  • Demonstrated antiproliferative activity in select cell lines (IC50 4.13 μM)
  • Molecular formula C23H29ClN8O2; molecular weight 484.98 g·mol⁻¹
  • Provided as a solid small-quantity package for research use
Catalog No. 50-003-08008