Learn More
Enzo Life Sciences SHPS-1/SIRP-1 alpha polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPTK140E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human SHPS-1/SIRP-1α. UniProt ID: P78324. Species reactivity: Human, Mouse, Rat. Applications: ICC, WB. Recommended Dilutions: Immunocytochemistry (1µg/ml) Western Blot (0.5-1.0µg/ml, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: DEAE purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7315