Learn More
MSC Value Collection Shrink Wrap, 20 in, 3500 ft, 1 Roll
Supplier:  MSC 39580311
Encompass
This Polyolefin Shrink Film offers excellent performance across manual, semi-automatic, and automatic equipment. It's a multi-layer, biaxially oriented film with high gloss, clarity, and puncture resistance. Suitable for cold storage and irregularly shaped items, it meets FDA food contact requirements and is U.S. Postal Service approved.
  • Requires no dwell time
  • Center-folded film
  • Works well on irregularly shaped items
Catalog No. 50-191-8441