Learn More
Medchemexpress LLC HY-P0227 1mg Medchemexpress, SHU 9119 CAS:168482-23-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02271MG
Encompass_Preferred
Medchemexpress, HY-P0227 1mg SHU 9119 CAS:168482-23-3 SHU 9119 is a potent human melanocortin 3 and 4 receptors ( MC3/4R ) antagonist and a partial MC5R agonist; with IC 50 values of 0.23, 0.06, and 0.09 nM for human MC3R, MC4R and MC5R, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4061