Learn More
Medchemexpress LLC SI-109 | 2429877-30-3 | 99.0% | 10 MG

Supplier: Medchemexpress LLC HY12960310MG
SI-109 is a potent STAT3 SH2 domain inhibitor (Ki=9 nM) that exhibits antitumor activity. It effectively inhibits the transcriptional activity of STAT3 (IC50=3 μM). This compound has been utilized, along with an analog of the CRBN ligand lenalidomide, to design the PROTAC STAT3 degrader SD-36.
- Potent STAT3 SH2 domain inhibitor
- Exhibits antitumor activity
- Inhibits transcriptional activity of STAT3
- Used in the design of PROTAC STAT3 degrader SD-36
- Exerts moderate growth inhibitory activity in MOLM-16 cells
- Solid, white to off-white appearance
- Shipped with dry ice
- Store at -80°C, under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.