Learn More
Medchemexpress LLC SI-109 | 2429877-30-3 | >99.91% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY129603
Encompass_Preferred
SI-109 is a potent inhibitor of the STAT3 SH2 domain, exhibiting antitumor activity by effectively inhibiting the transcriptional activity of STAT3. It has been utilized in the design of the PROTAC STAT3 degrader SD-36. This compound exerts moderate growth inhibitory activity in MOLM-16 cells. It is for research use only and not intended for human use.
  • Potent STAT3 SH2 domain inhibitor
  • Exhibits antitumor activity
  • Inhibits transcriptional activity of STAT3
  • Utilized in PROTAC STAT3 degrader design
  • Moderate growth inhibitory activity in MOLM-16 cells
Catalog No. 50-003-39193