Learn More
TARGETMOL CHEMICALS INC SI-113 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T958910MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. SI-113 is a potent and selective inhibitor of SGK1, a serine/threonine protein kinase, that modulates several oncogenic signaling cascades. Purity 98.81%
Catalog No. 50-287-5898