Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SI-2 hydrochloride | 1992052-49-9 | 98.1% | 301.77 g/mol | C15H16ClN5 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY101447A10MM 1ML

Encompass_Preferred

SI-2 hydrochloride (EPH 116 hydrochloride) is a small-molecule inhibitor of the steroid receptor coactivator-3 (SRC-3) protein-protein interaction used for research applications. The compound is provided as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL), and is characterized for in vitro and in vivo studies with reported purity and storage recommendations.

  • Potent SRC-3 inhibitor with low-nanomolar activity.
  • Supplied as solid and 10 mM solution in DMSO (1 mL).
  • High reported purity (98.06%).
  • Molecular weight approximately 301.77 g/mol.
  • Stable with recommended sealed storage and low-temperature solvent storage.

Catalog No. 50-002-48220


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.