Learn More
Medchemexpress LLC SI-2 hydrochloride | 1992052-49-9 | MFCD30290164 | 98.1% | 301.77 g/mol | C15H16ClN5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101447A25MG
Encompass_Preferred
SI-2 hydrochloride is the hydrochloride salt of a small-molecule SRC-3 protein-protein interaction inhibitor. It selectively reduces SRC-3 transcriptional activity and protein levels, induces breast cancer cell death at low nanomolar IC50 (3-20 nM), and has demonstrated tumor growth inhibition in MDA-MB-468 mouse models with improved toxicity and pharmacokinetic properties and acceptable oral availability.
  • Selective SRC-3 inhibitor with low-nanomolar cytotoxicity.
  • Demonstrated tumor growth inhibition in mouse models.
  • Improved toxicity and pharmacokinetic profile suitable for in vivo studies.
  • Soluble in DMSO at 5 mg/mL; recommends sealed, cold storage.
  • Stable in buffers at pH ≥ 5 with minimal short-term degradation at low pH.
  • Analytical purity approximately 98% (LCMS), suitable for research use.
Catalog No. 50-002-49069