Learn More
Medchemexpress LLC Si-W052 | 2572552-60-2 | 98.0% | 434.89 g·mol⁻¹ | C20H19ClN2O5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17406610MG
Encompass_Preferred
SI-W052 is an orally active, brain-penetrant small-molecule inhibitor that selectively targets mTOR and TEX264 to activate autophagy and promote endoplasmic reticulum turnover. It inhibits mTOR phosphorylation and reduces LPS-induced release of inflammatory cytokines, supporting research into neuroinflammation and Alzheimer's-associated pathways.
  • Orally active and brain-penetrant.
  • Selective inhibitor of mTOR and TEX264.
  • Activates autophagy via inhibition of mTOR phosphorylation.
  • Enhances TEX264 expression to promote endoplasmic reticulum turnover.
  • Reduces LPS-induced release of inflammatory cytokines such as TNF-α and IL-6.
  • Supplied as a high-purity solid with defined storage conditions for stability.
Catalog No. 50-002-99176