Learn More
Glass Expansion Sialon Injector for Axial D-Torch 2.4mm
SDP
Supplier:  Glass Expansion 318083469
Encompass_Preferred
Sialon Injector for Axial D-Torch 2.4mm. Compatible with Glass Expansion’s D-Torch for the Agilent (Varian) 700-ES and Vista Axial ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7226