Learn More
Cayman Chemical SIamycIn I 1mg
SDP
Supplier:  Cayman Chemical 277831 MG
Encompass_Preferred
A tricyclic peptide with antiviral and antibacterial activities; active against laboratory strains and clinical isolates of HIV-1 (ED50s = 0.05-0.45 and 0.89-5.7 μM, respectively), as well as the CBL-20 strain of HIV-2 (ED50 = 0.45 μM), in vitro; inhibits HIV-induced fusion of C8166 T cells with HIV-1-infected CEM-SS cells (ED50 = 0.08 μM); active against B. subtilis, M. luteus, and S. aureus (MICs = 1.6-6.3 μg/ml); inhibits GBAP-induced autophosphorylation of the E. faecalis quorum sensing kinase FsrC
Catalog No. 50-293-8683