Learn More
Carbosynth LLC. Siastatin B (5 mg vial), 5mg, MW: 218.21, Molecular formula: C8H14N2O5, CAS# 54795-58-3
SDP
Supplier:  Carbosynth LLC. CAR24002V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: (3S,4S,5R,6R)-6-(Acetylamino)-4,5-Dihydroxy-3-Piperidinecarboxylic Acid, Application: Microbial Product Inhibitor for Sialidase Glycosidase Inhibitors, Storage Temperature: 2-10C
Catalog No. 50-168-5728