Learn More
Vee Gee Vee Gee Glass Beaker 300 mL
Supplier:  Vee Gee 10020300A/DEL
Encompass_Preferred
The low-form Griffin beaker is made from high-quality borosilicate tubing and features an easy-pour spout and permanent white graduations. Uniform wall thickness - ideal for heating applications Made from high-quality borosilicate tubing with a low coefficient of expansion Graduation Accuracy: 5% of full capacity Graduation Range: 50 to 300 mL Graduation Interval: 50 mL
Catalog No. NC1937415
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.