Learn More
Medchemexpress LLC HY-12820 5mg Medchemexpress, Sibofimloc CAS:1616113-45-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128205MG
Encompass_Preferred
Medchemexpress, HY-12820 5mg Sibofimloc CAS:1616113-45-1 Sibofimloc (Antibiotic-202) is a first-in-class, gut-restricted, orally bioavailable FimH adhesion inhibitor extracted from patent WO2014100158A1, Compound Example 202. Sibofimloc has anti-bacterial infective activity and is developed for inflammatory bowel disease (IBD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1156