Learn More
Medchemexpress LLC Sid 26681509 | 958772-66-2 | 99.2% | 539.65 g/mol | C27H33N5O5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10335310MG
Encompass_Preferred
A potent, reversible, competitive small-molecule inhibitor of human cathepsin L with reported in vitro and in vivo activity. The compound is a white to off-white solid (molecular weight 539.65 g/mol, CAS 958772-66-2) and is supplied in small quantities for research use.
  • Potent cathepsin L inhibition (IC50 = 56 nM; potency increases to IC50 = 1.0 nM after 4 h preincubation).
  • High reported purity (99.16%).
  • Available as small solid quantities and as a 10 mM solution in DMSO for screening.
  • Demonstrated in vitro activity against Plasmodium falciparum and Leishmania major.
  • Suitable for biochemical assays and in vivo models of inflammation and sepsis.
Catalog No. 50-002-44576