Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC BOX Partners Box, 12.5 x 3 x 17.5 in, 25/Pk.

Supplier:  MSC 39554746

Encompass

This corrugated shipping box is designed for side loading, making it ideal for the easy insertion of flat products such as frames and mirrors. It features full overlap (FOL) flaps that provide enhanced protection for contents during transit.

  • Constructed with 1 wall for durability
  • White in color
  • Features a 65 lb capacity
  • Designed to ship flat, conserving storage space and reducing shipping costs
  • Made in USA

Catalog No. 50-191-5947


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.