Learn More
MSC Value Collection Side Mount Guidance Pulley, Malleable Iron, Galvanized, 1-1/8 in. Outside Dia, 1 ea
Supplier:  MSC 67328583
Encompass
This single side side mount guidance pulley is designed for use with fibrous rope. Constructed from durable malleable iron with a galvanized finish, this component is suited for various material handling and hardware applications.
  • Constructed from malleable iron
  • Features a galvanized finish
  • Designed for use with fibrous rope
  • Outside diameter of 1-1/8 in.
  • Mounting hole diameter of 0.157 in. to 0.168 in.
Catalog No. 50-005-12513