Learn More
Gilson Company Inc 3" Sieve, All Stainless, Full Height, No. 25
Supplier:  Gilson Company Inc V3SF #25
Encompass_Preferred
3in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 25 (710µm) openings and full height stainless steel frame. The overall height is 1-3/4in and the stacked height is 1-1/8in.
Catalog No. 50-871-303