Learn More
Gilson Company Inc 3in (76mm) diameter ASTM E11 Test Sieve, No. 45
Supplier:  Gilson Company Inc V3SF #45
Encompass_Preferred
3in (76mm) diameter ASTM E11 Test Sieve has stainless steel woven-wire cloth with No. 45 (355µm) openings and full height stainless steel frame. Overall height is 1-3/4in and stacked height is 1-1/8in. Features: Individually serial numbered, Compliant with ASTM E11 requirements for Compliance Sieves. Included Items: 3” Sieve, All Stainless, Full Height, No. 45, Certificate of Conformance to ASTM E11 requirements.
Catalog No. 50-871-307