Learn More
Gilson Company Inc SIEVE 3 SS HH NO3-1/2
Supplier:  Gilson Company Inc V3SH #31/2
Encompass_Preferred
Sieve, Test; Gilson; Fine series; Stainless-steel cloth/frame; Half height; 3 in. dia.; No. 3-1/2; Sieve opening; 5.6mm
Catalog No. 50-871-328