Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO4 -
Supplier:  Gilson Company Inc GAA21
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 4; Sieve opening; 4.75mm
Catalog No. 50-871-369