Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO50 -
Supplier:  Gilson Company Inc GAA37
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 50; Sieve opening; 300um
Catalog No. 50-871-385