Learn More
Gilson Company Inc SIEVE ACRYLIC 3 SS NO70 -
Supplier:  Gilson Company Inc GAA39
Encompass_Preferred
Sieve, Test; Gilson; For GilSonic AutoSiever; Acrylic frame with stainless-steel mesh; 3 in. dia.; No. 70; Sieve opening; 212um
Catalog No. 50-871-387