Learn More
Biodesign Inc of Ny Sieve Holder, two piece plastic with metal tightener,10 inches (25.4cm) diameter
Supplier:  Biodesign Inc of Ny F10
Encompass
Sieve Holder used to tightly hold CellMicroSieve filters to form a funnel through which extraction liquid can easily be collected
Catalog No. NC2684793